Learn More
Enzo Life Sciences Epidermal growth factor (mouse), (recombinant) (500µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2130500
Encompass_Preferred
Alternative name: Urogastrone, URG, EGF. Purity: ≥97% (SDS-PAGE & HPLC). Formulation: Lyophilized. Contains 0.1% trifluoroacetic acid. Source: Produced in E. coli. A non-glycoylated protein containnig 53 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4003