Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP548220UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Biotin. Host: E. coli. Species Reactivity: Mouse. BioActivity: 3T3 cell proliferation significantly induced by less than 250 pg/ml of Mouse EGF / Epidermal Growth Factor (which is equivalent to activity of at least 4.0 x 10^6 units/mg or an ED50 typically around 10-100 pg/ml). AA Sequence: MKKIDDDKNS . YPGCPSSYD G. YCLNGGVC MH. IESLDSY TCN. CVIGYS GDRC. QTRDL EWWEL. R. Buffer: 0.5 mg/mL in sterile PBS. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: EGF. Gene ID: 13645. Uniprot: P01132.
Catalog No. 50-188-5859