Learn More
Medchemexpress LLC HY-P7092 10ug Medchemexpress, EGF, Rat CAS: Purity:>98%
SDP
Catalog No. 501934330
Encompass_Preferred
Medchemexpress, HY-P7092 10ug EGF, Rat CAS: Recombinant Rat Epidermal Growth Factor is a potent epidermal growth factor, promotes epithelial proliferation and migration, and increases epithelial wound closure and shortens healing time. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4330 Supplier Medchemexpress LLC Supplier No. HYP709210UG
Medchemexpress, HY-P7092 10ug EGF, Rat CAS: Recombinant Rat Epidermal Growth Factor is a potent epidermal growth factor, promotes epithelial proliferation and migration, and increases epithelial wound closure and shortens healing time. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.