Learn More
Enzo Life Sciences Epidermal growth factor (rat), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX2018120100
Encompass_Preferred
Alternative name: EGF, Urogastrone, URG. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm). MW: ~6.3kDa. Formulation: Lyophilized from 10mM Na2PO4, pH 7.5. Sterile filtered. Source: Produced in E. coli. Non-glycosylated protein, containing 54 amino acids, with a molecular weight of 6.3 kDa. Long Term Storage: -20°C.
Catalog No. 50-200-8015