Learn More
Enzo Life Sciences EGF receptor (human) (652-657) (1mg)
SDP
Supplier:  Enzo Life Sciences ALX155023M001
Encompass_Preferred
Specific substrate (aa 652-657 of mature EGF receptor, without signal peptide (aa 1-24)) for protein kinase C (PKC). Alternative name: Epidermal growth factor receptor (652-657), EGFR (652-657), HER-1 (652-657). Formula: C34H68N16O8. MW: 829.0. Purity: ≥97%. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7812