Learn More
Enzo Life Sciences EGF receptor monoclonal antibody (6F1) (100µg)
SDP
Supplier:  Enzo Life Sciences ADICSA330E
Encompass_Preferred
Clone: 6F1. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant human EGF receptor (C-terminal fragment). UniProt ID: P00533. Species reactivity: Human. Applications: Flow Cytometry, ICC, IP, WB. Recommended Dilutions: Flow Cytometry (10µg/ml per 5x105 cells) Immunocytochemistry (10µg/ml) Immunoprecipitation (10-100µg, cell extract) Western Blot (1-5µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Lyophilized. In PBS, pH 7.2, containing 10mg/ml sucrose and 1mg/ml sodium azide. Reconstitute in 1 ml of distilled water.
Catalog No. 50-200-7183