Learn More
MilliporeSigma™ EGF-Stimulated A431 Cell Lysate
Catalog No. 12302MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100μ L
Catalog No. Quantity
12302MI 100μ L
1 options
Catalog No. 12302MI Supplier MilliporeSigma™ Supplier No. 12302
Add to Cart
Add to Cart
For use in Western Blotting
EGF-Stimulated A431 Cell Lysate in modified RIPA buffer (50mM Tris-HCl, pH 7.4, 1% NP40, 0.25% sodium deoxycholate, 150mM NaCl, 1mM EDTA, 1mM PMSF, 1mg/mL aprotinin, 1mg/mL leupeptin, 1mg/mL pepstatin, 1mM Na3VO4, 1mM NaF) and diluted with non-reducing sample buffer (31mM Tris-HCl, pH 6.8, 5% glycerol, 1% SDS, 0.002% bromophenol blue). Routinely evaluated as a positive control for many of Upstates antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 6 months at -20°C
Quantity 100μ L