Learn More
Medchemexpress LLC Egfp mrna-lnp | 500 UG
SDP
Catalog No. 5000372574
Encompass_Preferred
eGFP mRNA-LNP is a lipid nanoparticle (LNP) that contains eGFP mRNA. It carries Enhanced Green Fluorescent Protein (eGFP), which expresses green fluorescent protein upon entering a cell. eGFP is frequently utilized as a reporter gene, detectable by fluorescence microscopy or flow cytometry, with an excitation wavelength of 488 nm and an emission wavelength of 535/40 bandpass. This product can be employed for RNA delivery, assessing translation efficiency, and evaluating cell viability.
  • Contains eGFP mRNA.
  • Expresses green fluorescent protein upon entering the cell.
  • Used as a reporter gene.
  • Detectable by fluorescence microscopy or flow cytometry.
  • Employed for RNA delivery.
  • Assesses translation efficiency.
  • Evaluates cell viability.
Catalog No. 50-003-72574 Supplier Medchemexpress LLC Supplier No. HY153231A500UG
eGFP mRNA-LNP is a lipid nanoparticle (LNP) that contains eGFP mRNA. It carries Enhanced Green Fluorescent Protein (eGFP), which expresses green fluorescent protein upon entering a cell. eGFP is frequently utilized as a reporter gene, detectable by fluorescence microscopy or flow cytometry, with an excitation wavelength of 488 nm and an emission wavelength of 535/40 bandpass. This product can be employed for RNA delivery, assessing translation efficiency, and evaluating cell viability.
  • Contains eGFP mRNA.
  • Expresses green fluorescent protein upon entering the cell.
  • Used as a reporter gene.
  • Detectable by fluorescence microscopy or flow cytometry.
  • Employed for RNA delivery.
  • Assesses translation efficiency.
  • Evaluates cell viability.