Learn More
Cayman Chemical EGFP mRNACap-1mo5U 100ug
SDP
Catalog No. 502942107
Encompass_Preferred
An mRNA for the expression of EGFP; encapsulation in LNPs can be used for mRNA delivery and expression of EGFP in vitro or in vivo
Catalog No. 50-294-2107 Supplier Cayman Chemical Supplier No. 39800100 UG
An mRNA for the expression of EGFP; encapsulation in LNPs can be used for mRNA delivery and expression of EGFP in vitro or in vivo