Learn More
Abcam EGFP protein (His tag), 100UG
Supplier:  Abcam AB51992100UG
Encompass_Preferred
EGFP protein (His tag) is a Aequorea victoria Full Length protein, in the 2 to 238 aa range, expressed in Escherichia coli, with >=97% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, WB, Conjugation.
Protein/ Peptide/ Enzyme Synonyms: Green fluorescent protein, GFP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5232