Learn More
Apexbio Technology LLC EGFP saRNA (5mCTP) 100ug (1mg/mL)
SDP
Catalog No. 503107404
Encompass_Preferred
EGFP saRNA (5mCTP) encodes Enhanced green fluorescent protein (EGFP) a commonly used reporter molecule that emits strong and bright green fluorescence upon excitation with peak excitation/emission wavelengths at 488 nm/509 nm respectively Self-amplifying RNA (saRNA) due to its intracellular self-replication capability typically offers high expression levels prolonged duration and the advantage of low-dose usage EGFP saRNA provided at approximately 1 mg/mL concentration enables stable and prolonged expression of enhanced green fluorescent protein and can be used as a control to analyze efficiency in mRNA delivery and translation Incorporation of 5mCTP modifications suppresses RNA-mediated innate immune activation
Catalog No. 50-310-7404 Supplier Apexbio Technology LLC Supplier No. R30010.1
EGFP saRNA (5mCTP) encodes Enhanced green fluorescent protein (EGFP) a commonly used reporter molecule that emits strong and bright green fluorescence upon excitation with peak excitation/emission wavelengths at 488 nm/509 nm respectively Self-amplifying RNA (saRNA) due to its intracellular self-replication capability typically offers high expression levels prolonged duration and the advantage of low-dose usage EGFP saRNA provided at approximately 1 mg/mL concentration enables stable and prolonged expression of enhanced green fluorescent protein and can be used as a control to analyze efficiency in mRNA delivery and translation Incorporation of 5mCTP modifications suppresses RNA-mediated innate immune activation