Learn More
Biocare Medical, LLC Epidermal Growth Factor Receptor Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC ACI063CK
Encompass_Preferred
Epidermal Growth Factor Receptor antibody [H11] reacts with a 170kDa (wild type) and 145kDa (vIII variant) protein, identified as the first member of type I family of growth factor receptors (initially identified as EGR-Receptor). The EGFR antibody H11 clone shows no cross reactivity with c-erbB-2, cerbB-3 or c-erbB-4. Various studies have observed and reported over-expression of EGFR in tumors of breast (25%), brain, bladder, lung, gastric, esophagus, cervix, ovary and endometrium.
Catalog No. 50-824-75