Learn More
Carnabio Usa Inc EGFR/10UG/CYTOPLASMIC
Catalog No. 507540760
Encompass
EGFR, Cytoplasmic domain, Wild type, Amino Acid 669-1210, NP_005219.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0760 Supplier Carnabio Usa Inc Supplier No. 0811510
EGFR, Cytoplasmic domain, Wild type, Amino Acid 669-1210, NP_005219.2, Expressed in Insect (sf21); 10microg