Learn More
Enzo Life Sciences [pTyr992] EGF receptor (988-998) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP3230001
Encompass_Preferred
This sequence, DADEpYLIPQQG, is from an autophosphorylation site (Tyr992) of epidermal growth factor receptor (EGFR). An excellent substrate for mammalian PTP1B with Km=3.9 µM ) and Yersinia PTPase. May be used in non-radioactive phosphatase assays incorporating the BIOMOL GREENTM reagent. Alternative name: PTP1B substrate, [pTyr992] EGF receptor (988-998). Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1346