Learn More
Sino Biological Recombinant Human EGFR Protein (d752-759, GST Tag), Active
Catalog No. 502575170
Encompass_Preferred
Recombinant human EGFR (d752-759) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5170 Supplier Sino Biological Supplier No. E1012OG500
Recombinant human EGFR (d752-759) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.