Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1956MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: GFR450. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: A431 cells, Breast or Bladder cancer. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant extracellular domain of human EGFR protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~170kDa (wild type), ~145kDa (vIII variant).Chromosome: 7p11.2. Gene Symbol: EGFR. Gene ID: 1956. Ensemble: ENSG00000146648. Uniprot: NP_001333828.
Catalog No. 50-188-4659