Learn More
Medchemexpress LLC EGFR Canine sf9 H 100ug
SDP
Catalog No. 5000182786
Encompass_Preferred
EGFR Protein is a receptor tyrosine kinase that binds to ligands of the EGF family The high expression of EGFR protein can be used as a diagnostic marker for canine transitional cell carcinoma (TCC) EGFR Protein participate in the NF-kappa-B signaling pathway The high expression of EGFR protein can be used as a diagnostic marker for canine transitional cell carcinoma (TCC) EGFR Protein Canine (sf9 His) is the recombinant canine-derived EGFR protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-82786 Supplier Medchemexpress LLC Supplier No. HYP72986100UG
EGFR Protein is a receptor tyrosine kinase that binds to ligands of the EGF family The high expression of EGFR protein can be used as a diagnostic marker for canine transitional cell carcinoma (TCC) EGFR Protein participate in the NF-kappa-B signaling pathway The high expression of EGFR protein can be used as a diagnostic marker for canine transitional cell carcinoma (TCC) EGFR Protein Canine (sf9 His) is the recombinant canine-derived EGFR protein expressed by Sf9 insect cells with C-His labeled tag