Learn More
Description
- Gene name: EGFR
- Gene Alias: ERBB,ERBB1, HER1, PIG61, mENA
- Entrez GeneID: 1956
- Gene Description: Epidermal growth factor receptor (erythroblastic leukemia v
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. |
| Product Type | EGFR/CEN7q FISH Probe |
| Quantity | 200 μL |
For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.