Learn More
Carnabio Usa Inc EGFR(D746-750)/10UG/CYTOPLASM
Catalog No. 507540762
Encompass
EGFR [d746-750], Cytoplasmic domain, Mutant d746-750, Amino Acid 669-745, 751-1210(end), NP_005219.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0762 Supplier Carnabio Usa Inc Supplier No. 0852710
EGFR [d746-750], Cytoplasmic domain, Mutant d746-750, Amino Acid 669-745, 751-1210(end), NP_005219.2, Expressed in Insect (sf21); 10microg