Learn More
Sino Biological Recombinant Human EGFR Protein (C775S T790M L858R, GST Tag), Active
Catalog No. 502575285
Encompass_Preferred
Recombinant human EGFR (C775S T790M L858R) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5285 Supplier Sino Biological Supplier No. E10122D2G50
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human EGFR (C775S T790M L858R) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.