Learn More
Sino Biological Recombinant Human EGFR Protein (H773_V774insNPH, GST Tag)
Catalog No. 502575127
Encompass_Preferred
Recombinant human EGFR (H773_V774insNPH) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5127 Supplier Sino Biological Supplier No. E1013OG50
Recombinant human EGFR (H773_V774insNPH) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.