Learn More
Sino Biological Recombinant Human EGFR Protein (D761Y, GST Tag), Active
Catalog No. 502575167
Encompass_Preferred
Recombinant human EGFR (D761Y) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5167 Supplier Sino Biological Supplier No. E1012QG50
Recombinant human EGFR (D761Y) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.