Learn More
Medchemexpress LLC EGFR/ErbB-2/ErbB-4 inhibitor-2 | 179248-61-4 | 99.5% | 387.43 g·mol⁻1 | C23H21N3O3 | 5 MG
SDP
Catalog No. 5000263329
Encompass_Preferred
EGFR/ErbB-2/ErbB-4 inhibitor-2 (Compound 5) is a small-molecule research inhibitor of EGFR and ErbB family kinases. It demonstrates submicromolar potency versus EGFR and ErbB-2 and lower micromolar activity versus ErbB-4. The compound is supplied as a high-purity solid intended for biochemical and cell-based research; for research use only.
  • Submicromolar potency against EGFR and ErbB-2 (IC50 0.017 μM and 0.08 μM).
  • Active against ErbB-4 (IC50 1.91 μM).
  • High purity suitable for research (99.45%).
  • Supplied as an off-white to gray solid for convenient handling.
  • Available in multiple pack sizes including 5 MG.
  • Intended for biochemical and cell-based assays; not for human use.
Catalog No. 50-002-63329 Supplier Medchemexpress LLC Supplier No. HY1124205MG
Add to Cart
Add to Cart
EGFR/ErbB-2/ErbB-4 inhibitor-2 (Compound 5) is a small-molecule research inhibitor of EGFR and ErbB family kinases. It demonstrates submicromolar potency versus EGFR and ErbB-2 and lower micromolar activity versus ErbB-4. The compound is supplied as a high-purity solid intended for biochemical and cell-based research; for research use only.
  • Submicromolar potency against EGFR and ErbB-2 (IC50 0.017 μM and 0.08 μM).
  • Active against ErbB-4 (IC50 1.91 μM).
  • High purity suitable for research (99.45%).
  • Supplied as an off-white to gray solid for convenient handling.
  • Available in multiple pack sizes including 5 MG.
  • Intended for biochemical and cell-based assays; not for human use.