Learn More
Medchemexpress LLC Egfr/ErbB-2-IN-2 | 179248-61-4 | 99.45% | 387.43 | 10 MG
SDP
Catalog No. 5000437827
Encompass_Preferred
EGFR/ErbB-2-IN-2 is an inhibitor of EGFR and ErbB with IC50s of 0.017 μM, 0.08 μM, and 1.91 μM for EGFR, ErbB-2, and ErbB-4, respectively. It also inhibits c-Src, VEGFR2, and c-Fms with IC50s of 0.30, 2.2, and 14.0 μM, respectively.
  • It inhibits EGFR, ErbB-2, and ErbB-4.
  • It also inhibits c-Src, VEGFR2, and c-Fms.
  • It is for research use only.
Catalog No. 50-004-37827 Supplier Medchemexpress LLC Supplier No. HY11242010MG
EGFR/ErbB-2-IN-2 is an inhibitor of EGFR and ErbB with IC50s of 0.017 μM, 0.08 μM, and 1.91 μM for EGFR, ErbB-2, and ErbB-4, respectively. It also inhibits c-Src, VEGFR2, and c-Fms with IC50s of 0.30, 2.2, and 14.0 μM, respectively.
  • It inhibits EGFR, ErbB-2, and ErbB-4.
  • It also inhibits c-Src, VEGFR2, and c-Fms.
  • It is for research use only.