Learn More
Medchemexpress LLC EGFR/ErbB-2-IN-2 | 179248-61-4 | 99.5% | 387.43 | 100 MG
SDP
Catalog No. 5000438377
Encompass_Preferred
EGFR/ErbB-2-IN-2 is an EGFR and ErbB inhibitor with IC50s of 0.017 μM, 0.08 μM, and 1.91 μM for EGFR, ErbB-2, and ErbB-4 respectively. It also inhibits c-Src, VEGFR2, and c-Fms with IC50s of 0.30, 2.2, and 14.0 μM, respectively.
  • Egfr and ErbB inhibitor
  • Inhibits c-Src, VEGFR2, and c-Fms
  • Available in various quantities
  • Solution available in DMSO
  • Off-white to gray solid appearance
  • Research use only
Catalog No. 50-004-38377 Supplier Medchemexpress LLC Supplier No. HY112420100MG
Add to Cart
Add to Cart
EGFR/ErbB-2-IN-2 is an EGFR and ErbB inhibitor with IC50s of 0.017 μM, 0.08 μM, and 1.91 μM for EGFR, ErbB-2, and ErbB-4 respectively. It also inhibits c-Src, VEGFR2, and c-Fms with IC50s of 0.30, 2.2, and 14.0 μM, respectively.
  • Egfr and ErbB inhibitor
  • Inhibits c-Src, VEGFR2, and c-Fms
  • Available in various quantities
  • Solution available in DMSO
  • Off-white to gray solid appearance
  • Research use only