Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human EGFR (ErbB1), GST Tag Recombinant Protein

Catalog No. PR7295B
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7295B 100 μg
PV3872 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR7295B Supplier Invitrogen™ Supplier No. PR7295B
Add to Cart
Edge
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic Domain, Cytoplasmic.

EGFR, epidermal growth factor receptor, is a receptor tyrosine kinases that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. EGFR is encoded by the EGFR gene located on chromosome 7 in humans. EGFR belongs to the HER/ERbB family of proteins that includes three other receptor tyrosine kinases, ERbB2, ERbB3, ERbB4. EGFR is a transmembrane receptor and binding of its cognate ligands such as EGF (Epidermal Growth Factor) and TGF alpha (Transforming Growth Factor alpha) to the extracellular domain leads to EGFR dimerization followed by autophosphorylation of the tyrosine residues in the cytoplasmic domain. Overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00533
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.02% Triton X-100, 50% glycerol, 2mM DTT, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 1956
Molecular Weight (g/mol) 90.5 kDa
Name Human EGFR (ErbB1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2.7.10.1; 9030024J15Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; EC 2.7.10.1; egf receptor; Egfr; EGFR-related peptide; epidermal growth factor receptor; epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian); Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); ERBB; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; HER1; kinase EGFR; mENA; NISBD2; Oncogene ERBB; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; Urogastrone; wa2; wa-2; Wa5; waved 2
Common Name EGFR
Gene Symbol EGFR
Kinase Family EGF Receptor (ErbB) Kinase Family
Protein Length 668-1210
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 668-1210
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Tyrosine Kinases
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic Domain, Cytoplasmic
Form Liquid
Protein Subtype Receptor Tyrosine Kinases
Research Category Signal Transduction
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.