Learn More
Sino Biological Recombinant Human EGFR Protein (L747S, GST Tag), Active
Catalog No. 502575195
Encompass_Preferred
Recombinant human EGFR (L747S) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5195 Supplier Sino Biological Supplier No. E10122Y2G50
Recombinant human EGFR (L747S) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.