Learn More
Enzo Life Sciences EGF receptor (human) Recombinant monoclonal antibody (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4870200
Encompass_Preferred
Clone: Matuzumab. Isotype: IgGκ. Immunogen: Domain III of the extracellular domain of human EGFR. Species reactivity: Human. Applications: Flow Cytometry, WB, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3013