Learn More
Genscript Corporation EGFR/HER1 hFc Chimera Human
Catalog No. 5000058152
Encompass
The epidermal growth factor receptor is a transmembrane protein that is a receptor for members of the epidermal growth factor family of extracellular protein ligands The epidermal growth factor receptor is a member of the ErbB family of receptors a subfamily of four closely related receptor tyrosine kinases EGFR HER2/neu Her 3 and Her 4 Receptor tyrosine kinase binding ligands of the EGF family and activating several signaling cascades to convert extracellular cues into appropriate cellular responses
Catalog No. 50-000-58152 Supplier Genscript Corporation Supplier No. Z03919500
The epidermal growth factor receptor is a transmembrane protein that is a receptor for members of the epidermal growth factor family of extracellular protein ligands The epidermal growth factor receptor is a member of the ErbB family of receptors a subfamily of four closely related receptor tyrosine kinases EGFR HER2/neu Her 3 and Her 4 Receptor tyrosine kinase binding ligands of the EGF family and activating several signaling cascades to convert extracellular cues into appropriate cellular responses