Learn More
Medchemexpress LLC EGFR/HER2-IN-9 | 1637253-79-2 | C25H25ClFN5O4 | 5 MG
SDP
Catalog No. 5000372251
Encompass_Preferred
EGFR/HER2-IN-9 (Compound 1) is an EGFR and HER2 inhibitor for research use only.
  • Inhibits EGFR with an IC50 of 3.2 nM
  • Inhibits EGFR T790M with an IC50 of 8.3 nM
  • Inhibits HER2 with an IC50 of 14 nM
Catalog No. 50-003-72251 Supplier Medchemexpress LLC Supplier No. HY1531115MG
EGFR/HER2-IN-9 (Compound 1) is an EGFR and HER2 inhibitor for research use only.
  • Inhibits EGFR with an IC50 of 3.2 nM
  • Inhibits EGFR T790M with an IC50 of 8.3 nM
  • Inhibits HER2 with an IC50 of 14 nM