Learn More
Medchemexpress LLC HY-P7316 10ug Medchemexpress, EGFR, His (Sf9 insect cells-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083144
Encompass_Preferred
Medchemexpress, HY-P7316 10ug EGFR, His (Sf9 insect cells-expressed), Human CAS: Recombinant Human Epidermal Growth Factor Receptor, His (Sf9 insect cells-expressed) is a glycosylated polypeptide chain with the C-termimal 6×His Tag produced in Sf9 insect cells. EGFR, belongs to a family of tyrosine kinase receptors, plays important roles in cell growth and differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3144 Supplier Medchemexpress LLC Supplier No. HYP731610UG
Medchemexpress, HY-P7316 10ug EGFR, His (Sf9 insect cells-expressed), Human CAS: Recombinant Human Epidermal Growth Factor Receptor, His (Sf9 insect cells-expressed) is a glycosylated polypeptide chain with the C-termimal 6×His Tag produced in Sf9 insect cells. EGFR, belongs to a family of tyrosine kinase receptors, plays important roles in cell growth and differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.