Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human EGF R Protein, His Tag
Catalog No. 501956528
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 85-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cetuximab at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human EGF R, His Tag (Cat. No. EGR-HF2H5) with a linear range of 0.039-0.313 μg/mL (QC tested).
Catalog No. 50-195-6528 Supplier ACROBiosystems Supplier No. EGRHF2H5200UG
Recombinant Protein;200UG;FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 85-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cetuximab at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human EGF R, His Tag (Cat. No. EGR-HF2H5) with a linear range of 0.039-0.313 μg/mL (QC tested).