Learn More
Enzo Life Sciences EGF receptor (human) monoclonal antibody (13G8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804064C100
Encompass_Preferred
Clone: 13G8. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 1165-1186 of EGF receptor (epidermal growth factor receptor) conjugated to KLH. Species reactivity: Human, Mouse. Specificity: Recognizes the C-terminus of the EGF receptor (aa 1165-1186). Recognition is independent of phosphorylation of Tyr1173. Applications: ELISA, ICC, IP, WB Luminex. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9766