Learn More
Medchemexpress LLC EGFR Human HEK293 100ug | 100ug
SDP
Catalog No. 5000194441
Encompass_Preferred
The EGFR protein is a receptor tyrosine kinase that can bind to a variety of ligands such as EGF TGFA AREG epigen BTC epiregulin and HBEGF to initiate signaling cascades that mediate cellular responses This involves receptor dimerization autophosphorylation and recruitment of adapter proteins such as GRB2 activating downstream pathways such as RAS-RAF-MEK-ERK PI3-kinase-AKT PLCgamma-PKC and STAT EGFR Protein Human (HEK293 His-Avi) is the recombinant human-derived EGFR protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-94441 Supplier Medchemexpress LLC Supplier No. HYP78435100UG
The EGFR protein is a receptor tyrosine kinase that can bind to a variety of ligands such as EGF TGFA AREG epigen BTC epiregulin and HBEGF to initiate signaling cascades that mediate cellular responses This involves receptor dimerization autophosphorylation and recruitment of adapter proteins such as GRB2 activating downstream pathways such as RAS-RAF-MEK-ERK PI3-kinase-AKT PLCgamma-PKC and STAT EGFR Protein Human (HEK293 His-Avi) is the recombinant human-derived EGFR protein expressed by HEK293 with C-Avi C-His labeled tag