Learn More
Medchemexpress LLC Recombinant human epidermal growth factor receptor (EGFR) protein (T790M, L858R) | 10UG
SDP
Catalog No. 5000174111
Encompass_Preferred
Recombinant human epidermal growth factor receptor (EGFR) protein bearing the clinically relevant T790M and L858R mutations, expressed in Sf9 insect cells and supplied as a 0.22 μm filtered solution formulated in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, 5 mM DTT, and 0.1 M trehalose. The preparation is intended for biochemical and binding studies and has low endotoxin levels suitable for sensitive assays.
  • Recombinant human EGFR containing T790M and L858R mutations.
  • Expressed in Sf9 insect cells for eukaryotic post-translational processing.
  • Supplied as a filtered, stabilized protein solution for immediate use.
  • Formulated with glycerol and reducing agent to maintain stability.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
Catalog No. 50-001-74111 Supplier Medchemexpress LLC Supplier No. HYP70283210UG
Recombinant human epidermal growth factor receptor (EGFR) protein bearing the clinically relevant T790M and L858R mutations, expressed in Sf9 insect cells and supplied as a 0.22 μm filtered solution formulated in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, 5 mM DTT, and 0.1 M trehalose. The preparation is intended for biochemical and binding studies and has low endotoxin levels suitable for sensitive assays.
  • Recombinant human EGFR containing T790M and L858R mutations.
  • Expressed in Sf9 insect cells for eukaryotic post-translational processing.
  • Supplied as a filtered, stabilized protein solution for immediate use.
  • Formulated with glycerol and reducing agent to maintain stability.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.