Learn More
Medchemexpress LLC EGFR-IN-11 | 2463200-44-2 | 98.8% | 50 MG
SDP
Catalog No. 5000341123
Encompass_Preferred
EGFR-IN-11 is a fourth-generation EGFR-tyrosine kinase inhibitor (EGFR-TKI) with an IC50 of 18 nM for triple mutant EGFRL858R/T790M/C797S. It significantly suppresses EGFR phosphorylation, induces apoptosis, and arrests the cell cycle at G0/G1.
  • Suppresses EGFR phosphorylation
  • Induces apoptosis
  • Arrests cell cycle at G0/G1 phase
Catalog No. 50-003-41123 Supplier Medchemexpress LLC Supplier No. HY13061650MG
EGFR-IN-11 is a fourth-generation EGFR-tyrosine kinase inhibitor (EGFR-TKI) with an IC50 of 18 nM for triple mutant EGFRL858R/T790M/C797S. It significantly suppresses EGFR phosphorylation, induces apoptosis, and arrests the cell cycle at G0/G1.
  • Suppresses EGFR phosphorylation
  • Induces apoptosis
  • Arrests cell cycle at G0/G1 phase