Learn More
TARGETMOL CHEMICALS INC EGFR-IN-12 5MG
SDP
Catalog No. 502579800
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EGFR-IN-12 (EGFR Inhibitor) is a cell permeable and selective inhibitor of EGFR kinase (IC50 21 nM) and blocks receptor autophosphorylation in cells. purity: 99%
Catalog No. 50-257-9800 Supplier TARGETMOL CHEMICALS INC Supplier No. T51685MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EGFR-IN-12 (EGFR Inhibitor) is a cell permeable and selective inhibitor of EGFR kinase (IC50 21 nM) and blocks receptor autophosphorylation in cells. purity: 99%