Learn More
Medchemexpress LLC EGFR-IN-17 10mg | 2817679-31-3 | 568.01 | C27H31ClN7O3P | 10 MG
SDP
Catalog No. 5000226474
Encompass_Preferred
EGFR-IN-17 is a potent, selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR), active against resistance-associated mutants such as C797S, T790M, and L858R. Supplied as a 10 mg research-use compound, it is used to study EGFR signaling and mechanisms of drug resistance in biochemical and cellular assays.
  • Potent EGFR inhibition (IC50 0.0002 μM).
  • Active against resistance-associated mutants C797S, T790M, and L858R.
  • High purity suitable for research applications (98.18%).
  • Well-characterized chemical identity (MW 568.01; formula C27H31ClN7O3P).
  • Available in small quantities convenient for assay development.
  • Accompanied by a product data sheet with experimental and handling information.
Catalog No. 50-002-26474 Supplier Medchemexpress LLC Supplier No. HY11571610MG
EGFR-IN-17 is a potent, selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR), active against resistance-associated mutants such as C797S, T790M, and L858R. Supplied as a 10 mg research-use compound, it is used to study EGFR signaling and mechanisms of drug resistance in biochemical and cellular assays.
  • Potent EGFR inhibition (IC50 0.0002 μM).
  • Active against resistance-associated mutants C797S, T790M, and L858R.
  • High purity suitable for research applications (98.18%).
  • Well-characterized chemical identity (MW 568.01; formula C27H31ClN7O3P).
  • Available in small quantities convenient for assay development.
  • Accompanied by a product data sheet with experimental and handling information.