Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGFR-IN-17 25mg | 2817679-31-3 | 568.01 | C27H31ClN7O3P | 25 MG
SDP

Catalog No. 5000307850
Encompass_Preferred

EGFR-IN-17 is a potent, selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR) designed to overcome C797S-mediated resistance. It exhibits sub-nanomolar biochemical potency and is supplied as an off-white to light yellow solid for research use.

  • Potent EGFR inhibitor (IC50 0.0002 μM).
  • Active against EGFR mutants C797S, T790M, and L858R.
  • High reported purity (98.18%).
  • Molecular weight 568.01; chemical formula C27H31ClN7O3P.
  • Available in solid pack sizes including 25 mg.
  • Storage: powder -20°C (recommended long term) or 4°C (short term).
  • For research use only.

Catalog No. 50-003-07850 Supplier Medchemexpress LLC Supplier No. HY11571625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGFR-IN-17 is a potent, selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR) designed to overcome C797S-mediated resistance. It exhibits sub-nanomolar biochemical potency and is supplied as an off-white to light yellow solid for research use.

  • Potent EGFR inhibitor (IC50 0.0002 μM).
  • Active against EGFR mutants C797S, T790M, and L858R.
  • High reported purity (98.18%).
  • Molecular weight 568.01; chemical formula C27H31ClN7O3P.
  • Available in solid pack sizes including 25 mg.
  • Storage: powder -20°C (recommended long term) or 4°C (short term).
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.