Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGFR-IN-17 50mg | 2817679-31-3 | 568.01 | C27H31ClN7O3P | 50 MG
SDP

Catalog No. 5000307966
Encompass_Preferred

EGFR-IN-17 is a potent, selective inhibitor of the epidermal growth factor receptor (EGFR) with an IC50 of 0.0002 μM, supplied as a high-purity solid for in vitro research and compound reconstitution.

  • Potent EGFR inhibition (IC50 0.0002 μM).
  • High purity (98.18%).
  • Off-white to light yellow solid form.
  • Molecular weight 568.01 g/mol.
  • Soluble in DMSO at 12.5 mg/mL with warming and sonication.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes and as ready-to-use DMSO solutions for reconstitution.

Catalog No. 50-003-07966 Supplier Medchemexpress LLC Supplier No. HY11571650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGFR-IN-17 is a potent, selective inhibitor of the epidermal growth factor receptor (EGFR) with an IC50 of 0.0002 μM, supplied as a high-purity solid for in vitro research and compound reconstitution.

  • Potent EGFR inhibition (IC50 0.0002 μM).
  • High purity (98.18%).
  • Off-white to light yellow solid form.
  • Molecular weight 568.01 g/mol.
  • Soluble in DMSO at 12.5 mg/mL with warming and sonication.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes and as ready-to-use DMSO solutions for reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.