Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGFR-IN-17 5mg | 2817679-31-3 | 568.01 | C27H31ClN7O3P | 5 MG
SDP

Catalog No. 5000227542
Encompass_Preferred

EGFR-IN-17 is a potent, selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR) developed to overcome C797S-mediated resistance in mutant EGFR variants. It is intended for research use only and is supplied as a solid or as a DMSO solution for in vitro studies.

  • Potent EGFR inhibition (IC50 0.0002 μM).
  • Active against C797S, T790M, and L858R EGFR mutants.
  • Molecular formula C27H31ClN7O3P and molecular weight 568.01.
  • High purity (98.18%) and off-white to light yellow solid.
  • Available as small solid quantities (e.g., 5 MG) and as 10 mM solution in DMSO.
  • Soluble in DMSO (12.5 mg/mL); ultrasonic warming may be required.
  • Recommended storage: powder at -20 °C (up to 3 years); in solvent at -80 °C for long-term.

Catalog No. 50-002-27542 Supplier Medchemexpress LLC Supplier No. HY1157165MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGFR-IN-17 is a potent, selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR) developed to overcome C797S-mediated resistance in mutant EGFR variants. It is intended for research use only and is supplied as a solid or as a DMSO solution for in vitro studies.

  • Potent EGFR inhibition (IC50 0.0002 μM).
  • Active against C797S, T790M, and L858R EGFR mutants.
  • Molecular formula C27H31ClN7O3P and molecular weight 568.01.
  • High purity (98.18%) and off-white to light yellow solid.
  • Available as small solid quantities (e.g., 5 MG) and as 10 mM solution in DMSO.
  • Soluble in DMSO (12.5 mg/mL); ultrasonic warming may be required.
  • Recommended storage: powder at -20 °C (up to 3 years); in solvent at -80 °C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.