Learn More
Medchemexpress LLC Egfr-in-5 | 2225887-26-1 | 99.49% | 571.69 | 10 MG
SDP
Catalog No. 5000344015
Encompass_Preferred
EGFR-IN-5 is an EGFR inhibitor with IC50s of 10.4, 1.1, 34, and 7.2 nM for EGFR, EGFRL858R, EGFRL858R/T790M, and EGFRL858R/T790M/C797S, respectively.
  • Egfr inhibitor
  • Exhibits antiproliferative activity against human A549, HCC827, and NCI-H1975 cells
  • Solid, light yellow to yellow color
Catalog No. 50-003-44015 Supplier Medchemexpress LLC Supplier No. HY11141550MG
EGFR-IN-5 is an EGFR inhibitor with IC50s of 10.4, 1.1, 34, and 7.2 nM for EGFR, EGFRL858R, EGFRL858R/T790M, and EGFRL858R/T790M/C797S, respectively.
  • Egfr inhibitor
  • Exhibits antiproliferative activity against human A549, HCC827, and NCI-H1975 cells
  • Solid, light yellow to yellow color