Learn More
Medchemexpress LLC EGFR-IN-86 | 3055550-22-3 | 99.9% | 423.49 | 25 MG
SDP
Catalog No. 5000363965
Encompass_Preferred
EGFR-IN-86 is an EGFR inhibitor with an IC50 of 1.5 nM. It shows high activity against glioblastoma, inducing apoptosis and arresting U87 cell cycles in the G2/M phase. This product is for research use only and not sold to patients.
  • Potent EGFR inhibitor
  • High activity against glioblastoma
  • Induces apoptosis
  • Arrests U87 cell cycles in G2/M phase
Catalog No. 50-003-63965 Supplier Medchemexpress LLC Supplier No. 25MGHY149530
EGFR-IN-86 is an EGFR inhibitor with an IC50 of 1.5 nM. It shows high activity against glioblastoma, inducing apoptosis and arresting U87 cell cycles in the G2/M phase. This product is for research use only and not sold to patients.
  • Potent EGFR inhibitor
  • High activity against glioblastoma
  • Induces apoptosis
  • Arrests U87 cell cycles in G2/M phase