Learn More
Medchemexpress LLC EGFR-IN-86 | 3055550-22-3 | 99.9% | 423.49 | 5 MG
SDP
Catalog No. 5000360014
Encompass_Preferred
EGFR-IN-86 is an epidermal growth factor receptor (EGFR) inhibitor, characterized by its high activity against glioblastoma with an IC50 of 1.5 nM. This compound has demonstrated the ability to induce apoptosis and arrest the U87 cell cycle in the G2/M phase, indicating its potential as an anti-glioma agent.
  • Acts as an EGFR inhibitor with high potency.
  • Exhibits significant activity against glioblastoma.
  • Induces apoptosis in cells.
  • Arrests the U87 cell cycle in the G2/M phase.
  • Available in solid form, with a white to off-white appearance.
  • Purity of 99.92% ensures high quality for research applications.
  • Easily soluble in DMSO for in vitro studies.
  • Can be prepared for in vivo experiments using specific solvent protocols to ensure reliable results.
Catalog No. 50-003-60014 Supplier Medchemexpress LLC Supplier No. HY1495305MG
EGFR-IN-86 is an epidermal growth factor receptor (EGFR) inhibitor, characterized by its high activity against glioblastoma with an IC50 of 1.5 nM. This compound has demonstrated the ability to induce apoptosis and arrest the U87 cell cycle in the G2/M phase, indicating its potential as an anti-glioma agent.
  • Acts as an EGFR inhibitor with high potency.
  • Exhibits significant activity against glioblastoma.
  • Induces apoptosis in cells.
  • Arrests the U87 cell cycle in the G2/M phase.
  • Available in solid form, with a white to off-white appearance.
  • Purity of 99.92% ensures high quality for research applications.
  • Easily soluble in DMSO for in vitro studies.
  • Can be prepared for in vivo experiments using specific solvent protocols to ensure reliable results.