Learn More
TARGETMOL CHEMICALS INC EGFR-IN-9 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T111635MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EGFR-IN-9 is a potent EGFR kinase inhibitor with IC50s of 7 nM 28 nM for the wild type EGFR kinase and double mutant EGFR kinase (L858R/T790M). EGFR-IN-9 has antitumor activity. purity: 99%
Catalog No. 50-258-1557