Learn More
Apexbio Technology LLC EGFR Inhibitor 879127-07-8 1mg
SDP
Catalog No. 5000091473
Encompass_Preferred
EGFR Inhibitor (CAS 879127-07-8) is a cell-permeable pyrimidine small molecule that selectively targets the epidermal growth factor receptor (EGFR) tyrosine kinase By inhibiting EGFR kinase activity with an IC50 of 21 nM this compound disrupts receptor autophosphorylation and downstream signaling It also demonstrates inhibitory activity against EGFR mutants L858R and L861Q with IC50 values of 63 nM and 4 nM respectively Inhibition of EGFR leads to altered expression of apoptotic regulators promoting apoptosis in tumor cells EGFR Inhibitor is valuable for investigating EGFR-dependent cellular processes and mechanisms underlying cancer progression or resistance
Catalog No. 50-000-91473 Supplier Apexbio Technology LLC Supplier No. C33271
EGFR Inhibitor (CAS 879127-07-8) is a cell-permeable pyrimidine small molecule that selectively targets the epidermal growth factor receptor (EGFR) tyrosine kinase By inhibiting EGFR kinase activity with an IC50 of 21 nM this compound disrupts receptor autophosphorylation and downstream signaling It also demonstrates inhibitory activity against EGFR mutants L858R and L861Q with IC50 values of 63 nM and 4 nM respectively Inhibition of EGFR leads to altered expression of apoptotic regulators promoting apoptosis in tumor cells EGFR Inhibitor is valuable for investigating EGFR-dependent cellular processes and mechanisms underlying cancer progression or resistance