Learn More
Cayman Chemical EGFR InhIbItr 1mg
SDP
Catalog No. 502970284
Encompass_Preferred
A cell permeable, selective inhibitor of the EGFR kinase (IC50s = 21, 63, and 4 nM for EGFR and the L858R and L861Q EGFR mutants, respectively)
Catalog No. 50-297-0284 Supplier Cayman Chemical Supplier No. 153631 MG
Add to Cart
Add to Cart
A cell permeable, selective inhibitor of the EGFR kinase (IC50s = 21, 63, and 4 nM for EGFR and the L858R and L861Q EGFR mutants, respectively)