Learn More
Cayman Chemical EGFR InhIbItr 25mg
SDP
Catalog No. 502969153
Encompass_Preferred
A cell permeable, selective inhibitor of the EGFR kinase (IC50s = 21, 63, and 4 nM for EGFR and the L858R and L861Q EGFR mutants, respectively)
Catalog No. 50-296-9153 Supplier Cayman Chemical Supplier No. 1536325 MG
Add to Cart
Add to Cart
A cell permeable, selective inhibitor of the EGFR kinase (IC50s = 21, 63, and 4 nM for EGFR and the L858R and L861Q EGFR mutants, respectively)