Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGFR kinase inhibitor 3 | 2922402-03-5 | 99.0% | 575.64 g·mol⁻¹ | C31H25N7O3S | 10 MG
SDP

Catalog No. 5000288714
Encompass_Preferred

EGFR kinase inhibitor 3 is a bivalent ATP-allosteric inhibitor of the epidermal growth factor receptor (EGFR) intended for research use. It inhibits wild-type EGFR and clinically relevant activating and resistance mutations, and is supplied as a high-purity solid for laboratory studies.

  • Bivalent ATP-allosteric EGFR inhibitor with low-nanomolar potency.
  • Chemical formula: C31H25N7O3S.
  • Molecular weight: 575.64 g·mol⁻¹.
  • Purity: 99.03%.
  • Physical form: solid powder.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg.
  • CAS number: 2922402-03-5.

Catalog No. 50-002-88714 Supplier Medchemexpress LLC Supplier No. HY16229910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

EGFR kinase inhibitor 3 is a bivalent ATP-allosteric inhibitor of the epidermal growth factor receptor (EGFR) intended for research use. It inhibits wild-type EGFR and clinically relevant activating and resistance mutations, and is supplied as a high-purity solid for laboratory studies.

  • Bivalent ATP-allosteric EGFR inhibitor with low-nanomolar potency.
  • Chemical formula: C31H25N7O3S.
  • Molecular weight: 575.64 g·mol⁻¹.
  • Purity: 99.03%.
  • Physical form: solid powder.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg.
  • CAS number: 2922402-03-5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.