Learn More
Sino Biological Recombinant Human EGFR Protein (d746-750 L718Q T790M, GST Tag), Active
Supplier: Sino Biological E10122E3G50
Recombinant human EGFR (d746-750 L718Q T790M) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.